Learn More
Electron Microscopy Sciences Alconox Anionic Powder Detergent 4 LB
SDP
Catalog No. 50997492
Encompass_Preferred
The original anionic powder detergent for manual and ultrasonic cleaning. An odorless, high sequestering powder. It helps to decontaminate radioactive surfaces.
Catalog No. 50-997-492 Supplier Electron Microscopy Sciences Supplier No. 60510
The original anionic powder detergent for manual and ultrasonic cleaning. An odorless, high sequestering powder. It helps to decontaminate radioactive surfaces.