Learn More
Medchemexpress LLC Aniracetam | 72432-10-1 | MFCD00153767 | 99.9% | 219.24 | 10 MG
SDP
Catalog No. 5000345318
Encompass_Preferred
Aniracetam is an orally active neuroprotective agent with nootropic effects. It is used in research for cerebral dysfunctional disorders and can potentiate ionotropic quisqualate (iQA) responses in the CA1 region of rat hippocampal slices, as well as excitatory post synaptic potentials (EPSPs) in Schaffer collateral-commissural synapses. This agent can also prevent CO2-induced impairment of acquisition in hypercapnia model rats.
  • Orally active neuroprotective agent
  • Possesses nootropic effects
  • Potentiates ionotropic quisqualate responses
  • Enhances excitatory post synaptic potentials
  • Prevents CO2-induced impairment of acquisition
Catalog No. 50-003-45318 Supplier Medchemexpress LLC Supplier No. HY109321G
Aniracetam is an orally active neuroprotective agent with nootropic effects. It is used in research for cerebral dysfunctional disorders and can potentiate ionotropic quisqualate (iQA) responses in the CA1 region of rat hippocampal slices, as well as excitatory post synaptic potentials (EPSPs) in Schaffer collateral-commissural synapses. This agent can also prevent CO2-induced impairment of acquisition in hypercapnia model rats.
  • Orally active neuroprotective agent
  • Possesses nootropic effects
  • Potentiates ionotropic quisqualate responses
  • Enhances excitatory post synaptic potentials
  • Prevents CO2-induced impairment of acquisition