Learn More
Abcam Aniracetam, AMPA receptor poteniator, 250MG
Catalog No. 502707685
Encompass_Preferred
MW 219.24 Da, Purity >99%. Nootropic, positive allosteric modulator of AMPA receptors. Slows AMPA receptor deactivation and desensitisation. Antidpressant and anxiolytic in vivo. Also available in Kit: Ionotropic agonists (ab120323).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7685 Supplier Abcam Supplier No. AB120316250MG
MW 219.24 Da, Purity >99%. Nootropic, positive allosteric modulator of AMPA receptors. Slows AMPA receptor deactivation and desensitisation. Antidpressant and anxiolytic in vivo. Also available in Kit: Ionotropic agonists (ab120323).
The product is subject to the following: Abcam Restricted Use Statement