Learn More
Medchemexpress LLC Anisodamine (6-Hydroxyhyoscyamine) | 55869-99-3 | 305.37 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN05845MG
Encompass_Preferred
Anisodamine (6-Hydroxyhyoscyamine) is a belladonna alkaloid that acts as a non-subtype-selective muscarinic and nicotinic cholinoceptor antagonist. It is used in traditional Chinese medicine to improve microcirculation in states of shock and in organophosphate poisoning.
  • Non-subtype-selective muscarinic and nicotinic cholinoceptor antagonist
  • Used in traditional Chinese medicine
  • Improves microcirculation in shock states
  • Used in organophosphate poisoning
  • Targets MAChR and nicotinic cholinoceptors
  • Affects GPCR/G protein and neuronal signaling pathways
Catalog No. 50-003-88732