Learn More
Medchemexpress LLC Anisodamine hydrobromide | 55449-49-5 | 99.5% | 386.28 g/mol | C17H24BrNO4 | 10 MG
SDP
Catalog No. 5000297273
Encompass_Preferred
Anisodamine hydrobromide is the hydrobromide salt of anisodamine (6-hydroxyhyoscyamine), a belladonna alkaloid with muscarinic and nicotinic receptor activity. It is supplied as a high-purity research reagent for pharmacology and biochemical studies; not for clinical use.
  • High purity (approximately 99.5%).
  • Molecular weight 386.28 g/mol.
  • Hydrobromide salt increases water solubility for aqueous assays.
  • Supplied in small milligram quantities suitable for analytical and preclinical research.
  • Commonly used in muscarinic receptor pharmacology and biochemical research applications.
Catalog No. 50-002-97273 Supplier Medchemexpress LLC Supplier No. HYN0584A10MG
Anisodamine hydrobromide is the hydrobromide salt of anisodamine (6-hydroxyhyoscyamine), a belladonna alkaloid with muscarinic and nicotinic receptor activity. It is supplied as a high-purity research reagent for pharmacology and biochemical studies; not for clinical use.
  • High purity (approximately 99.5%).
  • Molecular weight 386.28 g/mol.
  • Hydrobromide salt increases water solubility for aqueous assays.
  • Supplied in small milligram quantities suitable for analytical and preclinical research.
  • Commonly used in muscarinic receptor pharmacology and biochemical research applications.