Learn More
Medchemexpress LLC Anisoin 100g
SDP
Catalog No. 5000204569
Encompass_Preferred
Anisoin (4 4 -Dimethoxybenzoin) is a pre-chromatographic fluorogenic labelling reagent for (liquid chromatography) analysis of guanidino compounds (Ex/Em 325/435 nm)[1]
Catalog No. 50-002-04569 Supplier Medchemexpress LLC Supplier No. HYW097777100G
Anisoin (4 4 -Dimethoxybenzoin) is a pre-chromatographic fluorogenic labelling reagent for (liquid chromatography) analysis of guanidino compounds (Ex/Em 325/435 nm)[1]