Learn More
Medchemexpress LLC Anisoin 500g
SDP
Supplier:  Medchemexpress LLC HYW097777500G
Encompass_Preferred
Anisoin (4 4 -Dimethoxybenzoin) is a pre-chromatographic fluorogenic labelling reagent for (liquid chromatography) analysis of guanidino compounds (Ex/Em 325/435 nm)[1]
Catalog No. 50-002-06013