Learn More
Medchemexpress LLC Anisoin 50g
SDP
Catalog No. 5000205554
Encompass_Preferred
Anisoin (4 4 -Dimethoxybenzoin) is a pre-chromatographic fluorogenic labelling reagent for (liquid chromatography) analysis of guanidino compounds (Ex/Em 325/435 nm)[1]
Catalog No. 50-002-05554 Supplier Medchemexpress LLC Supplier No. HYW09777750G
Anisoin (4 4 -Dimethoxybenzoin) is a pre-chromatographic fluorogenic labelling reagent for (liquid chromatography) analysis of guanidino compounds (Ex/Em 325/435 nm)[1]