Learn More
Medchemexpress LLC Anisole-piperazine-methanone-benzothiazole-p-methylpiperidine | 906258-69-3 | 99.1% | C25H30N4O2S | 1 ML
SDP
Catalog No. 5000353847
Encompass_Preferred
Anisole-piperazine-methanone-benzothiazole-p-methylpiperidine is an active compound intended for research use only. With a purity of 99.1% and a molecular formula of C25H30N4O2S, this compound appears as a white to off-white solid.
  • Active compound for research purposes
  • High purity of 99.1%
  • Soluble in DMSO at 16.67 mg/mL
  • Stable as powder at -20°C for 3 years or 4°C for 2 years
  • Stable in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-003-53847 Supplier Medchemexpress LLC Supplier No. HY14576610MM/1ML
Anisole-piperazine-methanone-benzothiazole-p-methylpiperidine is an active compound intended for research use only. With a purity of 99.1% and a molecular formula of C25H30N4O2S, this compound appears as a white to off-white solid.
  • Active compound for research purposes
  • High purity of 99.1%
  • Soluble in DMSO at 16.67 mg/mL
  • Stable as powder at -20°C for 3 years or 4°C for 2 years
  • Stable in solvent at -80°C for 6 months or -20°C for 1 month