Learn More
AG Scientific Inc Anisomycin 10 MG
Catalog No. 502268222
Encompass_Preferred
​​​​​​A​nisomycin (also known as Flagecidin) is isolated from several species of Streptomyces griseolus. It interferes with the synthesis of protein and DNA by inhibiting the peptidyl transferase of the 80S ribosomal system.Anisomycin is also a p38/JNK MAP activator, anti-protozoal agent, and an apoptosis promoter in promyelocytic leukemia cells, Jurkat cells, ventricular monocytes, and colon adenocarcinoma cells.Applications:Antibacterial, for its ability to reduce growth and block the diffusion of bacteriaAntiprotozoal, for its effectiveness in the destruction of protozoaInhibitor of nucleic acid and protein synthesis
Catalog No. 50-226-8222 Supplier AG Scientific Inc Supplier No. A104910MG
​​​​​​A​nisomycin (also known as Flagecidin) is isolated from several species of Streptomyces griseolus. It interferes with the synthesis of protein and DNA by inhibiting the peptidyl transferase of the 80S ribosomal system.Anisomycin is also a p38/JNK MAP activator, anti-protozoal agent, and an apoptosis promoter in promyelocytic leukemia cells, Jurkat cells, ventricular monocytes, and colon adenocarcinoma cells.Applications:Antibacterial, for its ability to reduce growth and block the diffusion of bacteriaAntiprotozoal, for its effectiveness in the destruction of protozoaInhibitor of nucleic acid and protein synthesis