Learn More
TARGETMOL CHEMICALS INC ANISOMYCIN 100MG
SDP
Catalog No. 502579290
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Anisomycin (NSC-76712) is an antibiotic isolated from various Streptomyces species. It interferes with protein and DNA synthesis by inhibiting peptidyl transferase or the 80S ribosome system. purity: 99%
Catalog No. 50-257-9290 Supplier TARGETMOL CHEMICALS INC Supplier No. T6758100MG
Also available in 10 mg 25 mg 50 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Anisomycin (NSC-76712) is an antibiotic isolated from various Streptomyces species. It interferes with protein and DNA synthesis by inhibiting peptidyl transferase or the 80S ribosome system. purity: 99%