Learn More
Medchemexpress LLC Anisomycin | 22862-76-6 | 265.31 | 5 MG
SDP
Catalog No. 5000385108
Encompass_Preferred
Anisomycin is a potent protein synthesis inhibitor which interferes with protein and DNA synthesis by inhibiting peptidyl transferase or the 80S ribosome system. It is also a JNK activator, increasing phospho-JNK, and functions as a bacterial antibiotic.
  • Potent protein synthesis inhibitor
  • Interferes with protein and DNA synthesis
  • Inhibits peptidyl transferase or the 80S ribosome system
  • JNK activator
  • Bacterial antibiotic
Catalog No. 50-003-85108 Supplier Medchemexpress LLC Supplier No. HY189825MG
Anisomycin is a potent protein synthesis inhibitor which interferes with protein and DNA synthesis by inhibiting peptidyl transferase or the 80S ribosome system. It is also a JNK activator, increasing phospho-JNK, and functions as a bacterial antibiotic.
  • Potent protein synthesis inhibitor
  • Interferes with protein and DNA synthesis
  • Inhibits peptidyl transferase or the 80S ribosome system
  • JNK activator
  • Bacterial antibiotic