Learn More
Medchemexpress LLC Anisomycin | 22862-76-6 | 99.5% | 265.31 | 50 MG
SDP
Catalog No. 5000385189
Encompass_Preferred
Anisomycin (Standard) is a potent protein synthesis inhibitor that interferes with protein and DNA synthesis by inhibiting peptidyl transferase or the 80S ribosome system. It also acts as a JNK activator, increasing phospho-JNK levels, and functions as a bacterial antibiotic. This analytical standard is intended for research and analytical applications, and is commonly used in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.
  • Potent protein synthesis inhibitor
  • Interferes with protein and DNA synthesis
  • Inhibits peptidyl transferase or the 80S ribosome system
  • JNK activator
  • Increases phospho-JNK levels
  • Bacterial antibiotic
  • Analytical standard for research and analytical applications
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-003-85189 Supplier Medchemexpress LLC Supplier No. HY18982R50MG
Anisomycin (Standard) is a potent protein synthesis inhibitor that interferes with protein and DNA synthesis by inhibiting peptidyl transferase or the 80S ribosome system. It also acts as a JNK activator, increasing phospho-JNK levels, and functions as a bacterial antibiotic. This analytical standard is intended for research and analytical applications, and is commonly used in qualitative, quantitative, and methodological research experiments involving techniques such as HPLC, GC, and MS.
  • Potent protein synthesis inhibitor
  • Interferes with protein and DNA synthesis
  • Inhibits peptidyl transferase or the 80S ribosome system
  • JNK activator
  • Increases phospho-JNK levels
  • Bacterial antibiotic
  • Analytical standard for research and analytical applications
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS