Learn More
Medchemexpress LLC Anisomycin | 22862-76-6 | 99.8% | 265.31 | 1 ML
SDP
Catalog No. 5000385285
Encompass_Preferred
Anisomycin is a potent protein synthesis inhibitor which interferes with protein and DNA synthesis by inhibiting peptidyl transferase or the 80S ribosome system. Anisomycin is a JNK activator, which increases phospho-JNK. Anisomycin is a bacterial antibiotic.
  • Potent protein synthesis inhibitor
  • Interferes with protein and DNA synthesis by inhibiting peptidyl transferase or the 80S ribosome system
  • JNK activator
  • Bacterial antibiotic
Catalog No. 50-003-85285 Supplier Medchemexpress LLC Supplier No. HY1898210MM/1ML
Anisomycin is a potent protein synthesis inhibitor which interferes with protein and DNA synthesis by inhibiting peptidyl transferase or the 80S ribosome system. Anisomycin is a JNK activator, which increases phospho-JNK. Anisomycin is a bacterial antibiotic.
  • Potent protein synthesis inhibitor
  • Interferes with protein and DNA synthesis by inhibiting peptidyl transferase or the 80S ribosome system
  • JNK activator
  • Bacterial antibiotic