Learn More
Medchemexpress LLC Anisomycin | 22862-76-6 | 99.8% | 265.31 | 100 MG
SDP
Catalog No. 5000385093
Encompass_Preferred
Anisomycin is a potent protein synthesis inhibitor that interferes with protein and DNA synthesis by inhibiting peptidyl transferase or the 80S ribosome system. It is also a JNK activator, which increases phospho-JNK, and a bacterial antibiotic.
  • Potent protein synthesis inhibitor
  • Interferes with protein and DNA synthesis
  • Inhibits peptidyl transferase or the 80S ribosome system
  • JNK activator
  • Bacterial antibiotic
Catalog No. 50-003-85093 Supplier Medchemexpress LLC Supplier No. HY18982R100MG
Anisomycin is a potent protein synthesis inhibitor that interferes with protein and DNA synthesis by inhibiting peptidyl transferase or the 80S ribosome system. It is also a JNK activator, which increases phospho-JNK, and a bacterial antibiotic.
  • Potent protein synthesis inhibitor
  • Interferes with protein and DNA synthesis
  • Inhibits peptidyl transferase or the 80S ribosome system
  • JNK activator
  • Bacterial antibiotic