Learn More
Medchemexpress LLC Anisomycin | 22862-76-6 | MFCD00077650 | 265.31 | C14H19NO4 | 10 MG
SDP
Catalog No. 5000295328
Encompass_Preferred
Anisomycin is a bacterial antibiotic and potent research reagent that inhibits peptidyl transferase activity of the 80S ribosome, blocking protein synthesis and activating stress-activated kinases (JNK). It is used in cell and biochemical assays to study translation inhibition, stress signaling, apoptosis, and effects on DNA/RNA synthesis. Recommended storage varies for powder and solution forms.
  • Potent protein synthesis inhibitor.
  • Activates JNK and stress signaling pathways.
  • Applicable to cell and biochemical assays for translation studies.
  • Documented CAS number for unambiguous identification.
  • Stable under recommended storage conditions for powder and solvent solutions.
Catalog No. 50-002-95328 Supplier Medchemexpress LLC Supplier No. HY18982R10MG
Anisomycin is a bacterial antibiotic and potent research reagent that inhibits peptidyl transferase activity of the 80S ribosome, blocking protein synthesis and activating stress-activated kinases (JNK). It is used in cell and biochemical assays to study translation inhibition, stress signaling, apoptosis, and effects on DNA/RNA synthesis. Recommended storage varies for powder and solution forms.
  • Potent protein synthesis inhibitor.
  • Activates JNK and stress signaling pathways.
  • Applicable to cell and biochemical assays for translation studies.
  • Documented CAS number for unambiguous identification.
  • Stable under recommended storage conditions for powder and solvent solutions.