Learn More
Medchemexpress LLC HY-18982 100mg , Anisomycin Flagecidin;Wuningmeisu C CAS:22862-76-6 Purity:98%
SDP
Catalog No. 501492832
Encompass_Preferred
Medchemexpress, HY-18982 100mg Anisomycin Flagecidin;Wuningmeisu C CAS:22862-76-6 Formula:C14H19NO4 JNK Purity:98% Anisomycin is a potent c-Jun N-terminal kinase ( JNK ) activator. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2832 Supplier Medchemexpress LLC Supplier No. HY18982100MG
Medchemexpress, HY-18982 100mg Anisomycin Flagecidin;Wuningmeisu C CAS:22862-76-6 Formula:C14H19NO4 JNK Purity:98% Anisomycin is a potent c-Jun N-terminal kinase ( JNK ) activator. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.