Learn More
Selleck Chemical LLC Anisomycin
SDP
Supplier:  Selleck Chemical LLC S7409200MG
Encompass_Preferred
Anisomycin (Flagecidin Wuningmeisu C) is a bacterial antibiotic isolated from Streptomyces griseolus which inhibits protein synthesis and also act as a JNK activator Anisomycin upregulates autophagy and increases apoptosis
Catalog No. 50-000-01417