Learn More
Selleck Chemical LLC Anisomycin
SDP
Catalog No. 5000001401
Encompass_Preferred
Anisomycin (Flagecidin Wuningmeisu C) is a bacterial antibiotic isolated from Streptomyces griseolus which inhibits protein synthesis and also act as a JNK activator Anisomycin upregulates autophagy and increases apoptosis
Catalog No. 50-000-01401 Supplier Selleck Chemical LLC Supplier No. S74091G
Anisomycin (Flagecidin Wuningmeisu C) is a bacterial antibiotic isolated from Streptomyces griseolus which inhibits protein synthesis and also act as a JNK activator Anisomycin upregulates autophagy and increases apoptosis