Learn More
Enzo Life Sciences Anisomycin (50 mg). CAS: 22862-76-6
SDP
Catalog No. 502012150
Encompass_Preferred
Antibiotic. Activates JNK/SAPKs, p38 and MAP kinases but not ERKs leading to the phosphorylation of S6, histone H3 and HMG-14 and the induction of the immediate-early genes c-fos and c-jun. Induces apoptosis in the human monoblastoid cell line. Used at 300 ng/ml. Inhibits peptidyl transferase at higher concentrations. Used in the eradication of bean mildew. Inhibits other pathogenic fungi in plants.  Alternative name: 2-(p-Methoxybenzyl)-3,4-pyrrolidinediol-3-acetate. Purity: ≥97% (TLC). Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2150 Supplier Enzo Life Sciences Supplier No. BMLST1020050
Antibiotic. Activates JNK/SAPKs, p38 and MAP kinases but not ERKs leading to the phosphorylation of S6, histone H3 and HMG-14 and the induction of the immediate-early genes c-fos and c-jun. Induces apoptosis in the human monoblastoid cell line. Used at 300 ng/ml. Inhibits peptidyl transferase at higher concentrations. Used in the eradication of bean mildew. Inhibits other pathogenic fungi in plants.  Alternative name: 2-(p-Methoxybenzyl)-3,4-pyrrolidinediol-3-acetate. Purity: ≥97% (TLC). Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: -20°C.