Learn More
TOKU-E ANISOMYCIN D SOLN 10X0.5ML
Catalog No. NC2068738
Encompass_Preferred
Antibiotic, ReadyMade™, sterile-filtered, 10 mg/ml in PBD (pH 4.6). Useful in cell culture, COVID-19 research.
Catalog No. NC2068738 Supplier TOKU-E Supplier No. A22610X0.5ML
Antibiotic, ReadyMade™, sterile-filtered, 10 mg/ml in PBD (pH 4.6). Useful in cell culture, COVID-19 research.