Learn More
Alkali Scientific Anisomycin from streptomyces griseolus [2 [(4 Methoxyphenyl)methyl] 3, 4 pyrrolidinediol 3 acetate], 100 Mg
SDP
Catalog No. 5000660749
Encompass_Preferred
Anisomycin is a potent inhibitor of protein synthesis commonly used in research to study cellular stress responses and gene regulation. It binds to the ribosomal 60S subunit, preventing peptide bond formation and effectively blocking translation. This inhibition also activates stress related signaling pathways, making it a valuable tool in molecular biology, neuroscience, and cancer research. It is frequently used to investigate apoptosis, transcriptional regulation, and intracellular signaling mechanisms. Supplied in a 100 mg quantity, it is suitable for multiple experimental applications requiring consistent and reproducible inhibition of protein synthesis.
Catalog No. 50-006-60749 Supplier Alkali Scientific Supplier No. ANI002CP
Anisomycin is a potent inhibitor of protein synthesis commonly used in research to study cellular stress responses and gene regulation. It binds to the ribosomal 60S subunit, preventing peptide bond formation and effectively blocking translation. This inhibition also activates stress related signaling pathways, making it a valuable tool in molecular biology, neuroscience, and cancer research. It is frequently used to investigate apoptosis, transcriptional regulation, and intracellular signaling mechanisms. Supplied in a 100 mg quantity, it is suitable for multiple experimental applications requiring consistent and reproducible inhibition of protein synthesis.