Learn More
Medchemexpress LLC Anisotropine (bromide) | 80-50-2 | 98.0% | 100 MG
SDP
Catalog No. 5000411897
Encompass_Preferred
Anisotropine (Octatropine) bromide is an orally active anticholinergic muscarinic antagonist. It can inhibit gastric acid secretion and is used as an adjunct to peptic ulcers.
  • Orally active anticholinergic muscarinic antagonist.
  • Inhibits gastric acid secretion.
  • Used as an adjunct to peptic ulcers.
Catalog No. 50-004-11897 Supplier Medchemexpress LLC Supplier No. HYB1475100MG
Anisotropine (Octatropine) bromide is an orally active anticholinergic muscarinic antagonist. It can inhibit gastric acid secretion and is used as an adjunct to peptic ulcers.
  • Orally active anticholinergic muscarinic antagonist.
  • Inhibits gastric acid secretion.
  • Used as an adjunct to peptic ulcers.