Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Anisotropine methyl bromide | 80-50-2 | MFCD00083234 | 98.0% | 362.35 g/mol | C17H32BrNO2 | 10 MG
SDP

Catalog No. 5000297987
Encompass_Preferred

Anisotropine bromide is a quaternary ammonium anticholinergic muscarinic antagonist supplied for laboratory research. It inhibits gastric acid secretion and is used as an adjunct in peptic ulcer studies. The reagent is provided as a powder with defined purity and storage recommendations.

  • Used as a muscarinic antagonist in research
  • Inhibits gastric acid secretion; used in peptic ulcer studies
  • Chemical formula C17H32BrNO2; molecular weight 362.35 g/mol
  • CAS number 80-50-2
  • Purity 98.0%
  • Powder storage: -20°C (3 years); in solvent: -80°C (6 months), -20°C (1 month)
  • Available pack sizes: 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg
  • For research use only; not for human or veterinary use
  • Datasheet available

Catalog No. 50-002-97987 Supplier Medchemexpress LLC Supplier No. HYB147510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Anisotropine bromide is a quaternary ammonium anticholinergic muscarinic antagonist supplied for laboratory research. It inhibits gastric acid secretion and is used as an adjunct in peptic ulcer studies. The reagent is provided as a powder with defined purity and storage recommendations.

  • Used as a muscarinic antagonist in research
  • Inhibits gastric acid secretion; used in peptic ulcer studies
  • Chemical formula C17H32BrNO2; molecular weight 362.35 g/mol
  • CAS number 80-50-2
  • Purity 98.0%
  • Powder storage: -20°C (3 years); in solvent: -80°C (6 months), -20°C (1 month)
  • Available pack sizes: 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg
  • For research use only; not for human or veterinary use
  • Datasheet available

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.