Learn More
TARGETMOL CHEMICALS INC ANISOTROPINE METHYLBROMI 100MG
SDP
Catalog No. 502576728
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Anisotropine Methylbromide is an anticholinergic agent and has been used for relief of gastrointestinal spasm and for the suppression of gastric acid secretion. purity: 99%
Catalog No. 50-257-6728 Supplier TARGETMOL CHEMICALS INC Supplier No. T4981100MG
Also available in 5 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Anisotropine Methylbromide is an anticholinergic agent and has been used for relief of gastrointestinal spasm and for the suppression of gastric acid secretion. purity: 99%