Learn More
Sigma Aldrich Fine Chemicals Biosciences Anisyl alcohol 98 fCC fG
Catalog No. 502903788
Encompass
Anisyl alcohol is a volatile aromatic compound mainly found in vanilla extracts and anise oil. It is used as a flavoring agent and fragrance ingredient.
Catalog No. 50-290-3788 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. W20990210KGK
Anisyl alcohol is a volatile aromatic compound mainly found in vanilla extracts and anise oil. It is used as a flavoring agent and fragrance ingredient.