Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Anisylacetone 100g | 104-20-1 | 100 G
SDP

Catalog No. 5000229735
Encompass_Preferred

Anisylacetone (p-methoxybenzylacetone) is a liquid research reagent used as a male fruit fly attractant and as a fragrance and flavor intermediate. It is supplied for laboratory research and reference use with documented purity and storage recommendations.

  • High purity: 99.40% (product page) and 99.87% (purity documentation).
  • Appearance: colorless to light yellow liquid; density 1.046 g/cm3.
  • Available as a solution (10 mM in DMSO, 1 mL) and bulk quantities (50 g, 100 g, larger by quote).
  • Storage recommendations: pure form -20°C (up to 3 years) or 4°C (up to 2 years); in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Intended for research use only; SDS and analytical documentation are provided.

Catalog No. 50-002-29735 Supplier Medchemexpress LLC Supplier No. HY116047100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Anisylacetone (p-methoxybenzylacetone) is a liquid research reagent used as a male fruit fly attractant and as a fragrance and flavor intermediate. It is supplied for laboratory research and reference use with documented purity and storage recommendations.

  • High purity: 99.40% (product page) and 99.87% (purity documentation).
  • Appearance: colorless to light yellow liquid; density 1.046 g/cm3.
  • Available as a solution (10 mM in DMSO, 1 mL) and bulk quantities (50 g, 100 g, larger by quote).
  • Storage recommendations: pure form -20°C (up to 3 years) or 4°C (up to 2 years); in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Intended for research use only; SDS and analytical documentation are provided.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.