Learn More
Medchemexpress LLC Anisylacetone 50g | 104-20-1 | 178.23 g/mol | C11H14O2 | 50 G
SDP
Catalog No. 5000229280
Encompass_Preferred
Anisylacetone (p-methoxybenzylacetone) is a small organic compound used as a male fruit-fly attractant and as an analytical standard for research and behavioral studies. It is a colorless to light-yellow liquid with formula C11H14O2 and a molecular weight of 178.23 g/mol, commonly supplied in laboratory package sizes such as 50 g.
  • Used as a male fruit-fly attractant
  • Intended for research and analytical applications
  • Colorless to light-yellow liquid (C11H14O2)
  • Molecular weight 178.23 g/mol
  • Available in bulk package sizes (for example, 50 g and 100 g)
  • Certificate of analysis provides purity and recommended storage conditions
Catalog No. 50-002-29280 Supplier Medchemexpress LLC Supplier No. HY11604750G
Add to Cart
Add to Cart
Anisylacetone (p-methoxybenzylacetone) is a small organic compound used as a male fruit-fly attractant and as an analytical standard for research and behavioral studies. It is a colorless to light-yellow liquid with formula C11H14O2 and a molecular weight of 178.23 g/mol, commonly supplied in laboratory package sizes such as 50 g.
  • Used as a male fruit-fly attractant
  • Intended for research and analytical applications
  • Colorless to light-yellow liquid (C11H14O2)
  • Molecular weight 178.23 g/mol
  • Available in bulk package sizes (for example, 50 g and 100 g)
  • Certificate of analysis provides purity and recommended storage conditions