Learn More
Leica Biosystems Anti-roll plate assembly for Leica blade holder, 70 mm, 100 μm
Catalog No. NC2159313
Encompass
Anti-roll plate assembly for cryostat microtomy that reduces section curling and helps produce consistently stretched sections for easier handling. It mounts to the blade holder and maintains a precise gap between the pressure plate and the section-stretching surface to improve section quality.
  • Prevents curling of frozen tissue sections during sectioning.
  • Fixed 70 mm width to support standard cryostat sections.
  • 100 μm spacing between pressure plate and section stretching plate for controlled section stretching.
  • Designed for mounting to compatible blade holders.
  • Supplied as an assembly for straightforward installation.
This item is not returnable. View return policy
Catalog No. NC2159313 Supplier Leica Biosystems Supplier No. 14047742492
Anti-roll plate assembly for cryostat microtomy that reduces section curling and helps produce consistently stretched sections for easier handling. It mounts to the blade holder and maintains a precise gap between the pressure plate and the section-stretching surface to improve section quality.
  • Prevents curling of frozen tissue sections during sectioning.
  • Fixed 70 mm width to support standard cryostat sections.
  • 100 μm spacing between pressure plate and section stretching plate for controlled section stretching.
  • Designed for mounting to compatible blade holders.
  • Supplied as an assembly for straightforward installation.