Learn More
Aviva Systems Biology Corp ANK1 Antibody - middle region (ARP41662_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41662_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANK1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ANK1. Alternate names: ANK, SPH1, SPH2ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57938