Learn More
Sino Biological Recombinant Human AKT2 Protein (Full Length, GST Tag)
Supplier: Sino Biological A1714G100
Recombinant full length human AKT2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.