Learn More
Aviva Systems Biology Corp ANKMY2 Antibody - N-terminal region (ARP57386_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57386_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANKMY2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ANKMY2. Alternate names: DKFZP564O043, ZMYND20ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Yeast
Catalog No. 50-014-54040