Learn More
Aviva Systems Biology Corp ANKRA2 Antibody - C-terminal region (ARP50675_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50675_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANKRA2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ANKRA2. Alternate names: ANKRAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60408