Learn More
Aviva Systems Biology Corp ANKRD11 Antibody - N-terminal region (ARP33542_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33542_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ANKRD11. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ANKRD11. Alternate names: T13, LZ16, ANCO1, ANCO-1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55062