Learn More
Aviva Systems Biology Corp ANKRD37 Antibody - middle region (ARP55804_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55804_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANKRD37. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ANKRD37. Alternate names: Lrp2bp, MGC111507ApplicationWB. Species reactivity: Dog, Goat, Horse, Human
Catalog No. 50-014-61620