Learn More
Aviva Systems Biology Corp ANKRD39 Antibody - N-terminal region (ARP56912_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56912_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANKRD39. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ANKRD39. Alternate names: MGC41816ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-235