Learn More
Aviva Systems Biology Corp ANKRD42 Antibody - N-terminal region (ARP41193_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41193_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANKRD42. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ANKRD42. Alternate names: FLJ37874, SARPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-63002