Learn More
Aviva Systems Biology Corp ANKRD47 Antibody - N-terminal region (ARP55871_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55871_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANKRD47. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ANKRD47. Alternate names: FLJ46061, ANKRD47ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61640