Learn More
Aviva Systems Biology Corp ANKRD5 Antibody - middle region (ARP57592_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57592_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANKRD5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ANKRD5. Alternate names: FLJ21669, dJ839B4.6, ANKRD5ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-63024