Learn More
Aviva Systems Biology Corp ANKRD5 Antibody - N-terminal region (ARP57591_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57591_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANKRD5. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ANKRD5. Alternate names: FLJ21669, dJ839B4.6, ANKRD5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-337