Learn More
Aviva Systems Biology Corp Ankrd50 Antibody - C-terminal region (ARP57388_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57388_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ankrd50. It was validated on Western Blot. . Alternate names: RGD1311665ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-304