Learn More
Aviva Systems Biology Corp ANKRD7 Antibody - middle region (ARP53739_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53739_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ANKRD7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ANKRD7. Alternate names: TSA806ApplicationWB. Species reactivity: Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61112