Learn More
Aviva Systems Biology Corp Ankrd9 Antibody - C-terminal region (ARP52889_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52889_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ankrd9. It was validated on Western Blot. . Alternate names: 2500003O20RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-285-782