Learn More
Aviva Systems Biology Corp Anks1 Antibody - C-terminal region (ARP55208_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55208_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Anks1. It was validated on Western Blot. . Alternate names: Anks1a, MGC59413, mKIAA0229ApplicationIP, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-015