Learn More
Foxx Life Sciences VersaCap 83mm (83B) Carboy and Bottle Cap Adapters, Tangle Free Hose Barb Adapter, Two 1/2 Inch Barbs for 1/2 Inch ID Tubing, for use with VersaCap 83mm Open Cap
Catalog No. 501049838
Encompass_Preferred
205-5002-RLS: Adapter, 83mm, 2x 1/2" Molded In HB. Patented VersaCap is leak-proof, uses a replaceable Pt-cured silicone gasket, and provides the ultimate in top-connection flexibility. Open VersaCap is compatible with a variety of adapter inserts, enabling users to quickly change the top connections of each carboy or bottle to match their application needs. VersaCap offers large finger grips that allow for easy tightening and opening. They are also stackable, and offered in 53mm, 83mm (compatible with most carboys), and 120mm wide-mouth versions that fit exclusively on Foxx Life Sciences 20L, 40L, and 75L EZfil carboys. Quick-connect hose barb options include combinations of two, three, and four ports, with 1/4 and 3/8 hose barb sizes. Molded-in hose barb options are available in 1/8, 1/4, 516, and 1/2 sizes. VersaCap are made from polypropylene, which may be autoclaved at 15 PSI and 121C (250F) for 20 minutes.
Catalog No. 50-104-9838 Supplier Foxx Life Sciences Supplier No. 2055002RLS
205-5002-RLS: Adapter, 83mm, 2x 1/2" Molded In HB. Patented VersaCap is leak-proof, uses a replaceable Pt-cured silicone gasket, and provides the ultimate in top-connection flexibility. Open VersaCap is compatible with a variety of adapter inserts, enabling users to quickly change the top connections of each carboy or bottle to match their application needs. VersaCap offers large finger grips that allow for easy tightening and opening. They are also stackable, and offered in 53mm, 83mm (compatible with most carboys), and 120mm wide-mouth versions that fit exclusively on Foxx Life Sciences 20L, 40L, and 75L EZfil carboys. Quick-connect hose barb options include combinations of two, three, and four ports, with 1/4 and 3/8 hose barb sizes. Molded-in hose barb options are available in 1/8, 1/4, 516, and 1/2 sizes. VersaCap are made from polypropylene, which may be autoclaved at 15 PSI and 121C (250F) for 20 minutes.