Learn More
Hach Company Anlg Cond Sens Inpt Mod SC200
Catalog No. 5000207434 Shop All Hach Company Products
Encompass
Analog Conductivity Sensor Input Module for SC200 Universal ControllerThe Analog Conductivity module for the SC200 is a user installed module that allows for connection of analog conductivity sensors to the controller This module works with both contacting and inductive analog conductivity sensors See manual for additional connection information
Catalog No. 50-002-07434 Supplier Hach Company Supplier No. 9013000
Analog Conductivity Sensor Input Module for SC200 Universal ControllerThe Analog Conductivity module for the SC200 is a user installed module that allows for connection of analog conductivity sensors to the controller This module works with both contacting and inductive analog conductivity sensors See manual for additional connection information