Learn More
Selleck Chemical LLC Anlotinib (AL3818) dihydrochloride 5mg 1360460-82-7
SDP
Catalog No. 501948067
Encompass_Preferred
Anlotinib (AL3818) is a highly potent and selective VEGFR2 inhibitor with IC50 less than 1 nM. It has broad-spectrum antitumor potential in clinical trials. Please use saline solution rather than PBS for dilutions. PBS may cause precipitation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8067 Supplier Selleck Chemical LLC Supplier No. S87265MG
Anlotinib (AL3818) is a highly potent and selective VEGFR2 inhibitor with IC50 less than 1 nM. It has broad-spectrum antitumor potential in clinical trials. Please use saline solution rather than PBS for dilutions. PBS may cause precipitation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.