Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Annexin A1/ANXA1 Mo 100ug
SDP

Catalog No. 5000196271
Encompass_Preferred

Recombinant mouse Annexin A1 (ANXA1) is supplied lyophilized as a His-tagged protein expressed in E. coli and intended for biochemical and cell-based assays. ANXA1 is involved in immune regulation and exhibits anti-inflammatory activity; functional activity is demonstrated by inhibition of TNF-α secretion from RAW264.7 mouse macrophage cells.

  • Recombinant mouse Annexin A1 protein, suitable for biochemical and cell-based assays.
  • N-terminal 6xHis tag facilitates purification and detection.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Approximate molecular weight ~40 kDa.
  • Lyophilized formulation with trehalose for improved stability.
  • Storage at -20°C for up to 2 years; aliquot and store at -80°C for extended storage.
  • Activity validated by inhibition of TNF-α secretion in RAW264.7 cells.

Catalog No. 50-001-96271 Supplier Medchemexpress LLC Supplier No. HYP72078100UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant mouse Annexin A1 (ANXA1) is supplied lyophilized as a His-tagged protein expressed in E. coli and intended for biochemical and cell-based assays. ANXA1 is involved in immune regulation and exhibits anti-inflammatory activity; functional activity is demonstrated by inhibition of TNF-α secretion from RAW264.7 mouse macrophage cells.

  • Recombinant mouse Annexin A1 protein, suitable for biochemical and cell-based assays.
  • N-terminal 6xHis tag facilitates purification and detection.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Approximate molecular weight ~40 kDa.
  • Lyophilized formulation with trehalose for improved stability.
  • Storage at -20°C for up to 2 years; aliquot and store at -80°C for extended storage.
  • Activity validated by inhibition of TNF-α secretion in RAW264.7 cells.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.