Learn More
Medchemexpress LLC Annexin A1/ANXA1 Mo 100ug
SDP
Supplier:  Medchemexpress LLC HYP71713100UG
Encompass_Preferred
Recombinant full-length mouse Annexin A1 supplied lyophilized for research applications. The protein is expressed in Pichia pastoris with an N-terminal His tag, formulated in PBS with trehalose to improve stability, and provided at a high purity suitable for biochemical characterization and antibody generation. Handle as a frozen reagent and aliquot after reconstitution to preserve activity.
  • Full-length mouse protein with N-terminal His tag.
  • Expressed in Pichia pastoris for eukaryotic processing.
  • Lyophilized formulation in PBS with 6% trehalose for enhanced stability.
  • High purity (>90%) verified by reducing SDS-PAGE.
  • Suitable for biochemical assays, antibody production, and functional studies.
  • Store frozen and aliquot to avoid repeated freeze-thaw cycles.
Catalog No. 50-001-95637