Learn More
Medchemexpress LLC Annexin A1/ANXA1 Mo 10ug
SDP
Catalog No. 5000195542
Encompass_Preferred
Recombinant mouse Annexin A1 (His-tagged) is a full-length protein expressed in E. coli, provided lyophilized and quality-tested for purity and biological activity. It is designed for in vitro biochemical characterization and cell-based assays that require a His-tagged mouse Annexin A1 protein.
  • Full-length mouse protein with N-terminal 6xHis tag.
  • Expressed in E. coli and >95% pure by reducing SDS-PAGE.
  • Lyophilized formulation with trehalose for improved stability.
  • Demonstrated biological activity: inhibits TNF-alpha secretion in RAW264.7 cells.
  • Stable when stored at -20°C; aliquot and store at -80°C for extended storage.
Catalog No. 50-001-95542 Supplier Medchemexpress LLC Supplier No. HYP7207810UG
Recombinant mouse Annexin A1 (His-tagged) is a full-length protein expressed in E. coli, provided lyophilized and quality-tested for purity and biological activity. It is designed for in vitro biochemical characterization and cell-based assays that require a His-tagged mouse Annexin A1 protein.
  • Full-length mouse protein with N-terminal 6xHis tag.
  • Expressed in E. coli and >95% pure by reducing SDS-PAGE.
  • Lyophilized formulation with trehalose for improved stability.
  • Demonstrated biological activity: inhibits TNF-alpha secretion in RAW264.7 cells.
  • Stable when stored at -20°C; aliquot and store at -80°C for extended storage.